Learn More
Med Vet International McKesson Sterile USP Type VII Gauze Sponge, 16-Ply, 4 x 4 Inch, 16-4276, 10/Tray
Catalog No. 503044062
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 16-Ply 4 X 4 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 10 gauze sponges in a sterile tray 4 x 4 inches
Catalog No. 50-304-4062 Supplier Med Vet International Supplier No. 474706TR
USP Type VII Gauze Sponge McKesson Cotton 16-Ply 4 X 4 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 10 gauze sponges in a sterile tray 4 x 4 inches