Learn More
Med Vet International Cypress NonSterile Gauze Sponge, 8-Ply, 2 x 2 Inch, 40-28, 200/Box
Catalog No. 503045101
Encompass_Preferred
Gauze Sponge McKesson Brand Cotton 8-Ply 2 X 2 Inch Square NonSterile 8 ply Nonsterile Folded edges for reduced lint and loose threads High quality cotton yarn for enhanced absorbency and purity 2 x 2 inches
Catalog No. 50-304-5101 Supplier Med Vet International Supplier No. 867172BX
Gauze Sponge McKesson Brand Cotton 8-Ply 2 X 2 Inch Square NonSterile 8 ply Nonsterile Folded edges for reduced lint and loose threads High quality cotton yarn for enhanced absorbency and purity 2 x 2 inches