Learn More
Med Vet International Dynarex Surgical Gauze Sponge, Sterile 2'S, 4" X 4", 12 Ply, 600/Case
Catalog No. 502829271
Encompass_Preferred
Gauze sponges and pads are ideal for general cleaning, dressing and packing wounds. Dynarex X-Ray Detectable Gauze Sponges are economical, multi-purpose gauze sponges ideal for use in surgical procedures. Highly absorbent, 100% cotton gauze Folded edges minimizes loose threads and lint 3351 - Blue radiopaque element for easy recognition under X-ray; Not made with natural rubber latex Packaged in peel-down pouches for convenient, aseptic handling Packed in open dispenser boxes for convenient handling
Catalog No. 50-282-9271 Supplier Med Vet International Supplier No. 3343
Gauze sponges and pads are ideal for general cleaning, dressing and packing wounds. Dynarex X-Ray Detectable Gauze Sponges are economical, multi-purpose gauze sponges ideal for use in surgical procedures. Highly absorbent, 100% cotton gauze Folded edges minimizes loose threads and lint 3351 - Blue radiopaque element for easy recognition under X-ray; Not made with natural rubber latex Packaged in peel-down pouches for convenient, aseptic handling Packed in open dispenser boxes for convenient handling