Learn More
Med Vet International McKesson Sterile USP Type VII Gauze Sponge, 8-Ply, 2 x 2 Inch, 16-42228, 100/Box
Catalog No. 503043422
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 2 x 2 inches
Catalog No. 50-304-3422 Supplier Med Vet International Supplier No. 460859BX
USP Type VII Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 2 x 2 inches