Learn More
Med Vet International McKesson NonSterile Gauze Sponge, 8-Ply, 2 x 2 Inch, 22082000, 200/Box
Catalog No. 503044866
Encompass_Preferred
Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 2 x 2 inches
Catalog No. 50-304-4866 Supplier Med Vet International Supplier No. 373772BX
Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square NonSterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Nonsterile gauze sponge offers reduced linting properties Intended for single-use applications 2 x 2 inches