Learn More
Med Vet International McKesson Sterile Gauze Sponge, 8-Ply, 2 x 2 Inch, 16-4228, 3000/Case
Catalog No. 503043672
Encompass_Preferred
Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square Sterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Sterile gauze sponge offers reduced linting properties Intended for single-use applications Peelable pouch simplifies access Two woven gauze sponges per pack, 50 packs per box 2 x 2 inches
Catalog No. 50-304-3672 Supplier Med Vet International Supplier No. 635942CS
Gauze Sponge McKesson Cotton 8-Ply 2 X 2 Inch Square Sterile 100% cotton is ideal for wound dressing, cleaning, prepping and packing Sterile gauze sponge offers reduced linting properties Intended for single-use applications Peelable pouch simplifies access Two woven gauze sponges per pack, 50 packs per box 2 x 2 inches