Learn More
Med Vet International McKesson Sterile USP Type VII Gauze Sponge, 8-Ply, 4 x 4 Inch, 16-42448, 100/Box
Catalog No. 503044705
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 8-Ply 4 X 4 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches
Catalog No. 50-304-4705 Supplier Med Vet International Supplier No. 447085BX
USP Type VII Gauze Sponge McKesson Cotton 8-Ply 4 X 4 Inch Square Sterile Made from high-grade cotton of USP Type VII gauze for wound dressing, cleaning, prepping or packing 100% cotton construction offers reduced linting Intended for single-use applications USP means gauze sponge meets United States Pharmacopeial Convention standards 4 x 4 inches