Learn More
Med Vet International Nonwoven Sponge Curity™ 2 X 2 Inch, Sterile 4-Ply Square, 8042, 2/Pack
Catalog No. 503044639
Encompass_Preferred
4 ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 2 x 2 inches
Catalog No. 50-304-4639 Supplier Med Vet International Supplier No. 529380PK
4 ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 2 x 2 inches