Learn More
Med Vet International Dynarex Surgical Gauze Sponge - Sterile 10'S, 4" X 4" 12 Ply, 1280/Case
Catalog No. 502829263
Encompass_Preferred
Dynarex Gauze Sponges & Pads are premium-quality dressings crafted from highly absorbent 100% cotton materials, ideal for a variety of medical uses, including cleaning, dressing, and packing wounds. The Sponges & Pads feature folded edges to help minimize loose threads and lint. Dynarex Gauze Sponges & Pads are available in sterile, non-sterile, and x-ray-detectable varieties. Sterile varieties are packaged in peel-down pouches for convenient, aseptic handling. Dynarex Gauze Sponges & Pads are high-quality dressings made with 100% cotton materials for use in cleaning, dressing, and packing wounds. 
Catalog No. 50-282-9263 Supplier Med Vet International Supplier No. 3345
Dynarex Gauze Sponges & Pads are premium-quality dressings crafted from highly absorbent 100% cotton materials, ideal for a variety of medical uses, including cleaning, dressing, and packing wounds. The Sponges & Pads feature folded edges to help minimize loose threads and lint. Dynarex Gauze Sponges & Pads are available in sterile, non-sterile, and x-ray-detectable varieties. Sterile varieties are packaged in peel-down pouches for convenient, aseptic handling. Dynarex Gauze Sponges & Pads are high-quality dressings made with 100% cotton materials for use in cleaning, dressing, and packing wounds. 