Learn More
Med Vet International McKesson Sterile USP Type VII Gauze Sponge, 12-Ply, 3 x 3 Inch, 16-42332, 80/Box
Catalog No. 503044702
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 3 X 3 Inch Square Sterile They're made from 100% high-grade cotton for comfort These 3 x 3-inch USP Type VII gauze are suitable for dressing, cleaning, prepping or packing wounds They create a low amount of lint to keep the wound area clean These McKesson woven gauze sponges are sterile to help ward off infection Each 12-ply sponge comes in a peelable pouch for easy access
Catalog No. 50-304-4702 Supplier Med Vet International Supplier No. 447084BX
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 3 X 3 Inch Square Sterile They're made from 100% high-grade cotton for comfort These 3 x 3-inch USP Type VII gauze are suitable for dressing, cleaning, prepping or packing wounds They create a low amount of lint to keep the wound area clean These McKesson woven gauze sponges are sterile to help ward off infection Each 12-ply sponge comes in a peelable pouch for easy access