Learn More
Med Vet International McKesson Sterile USP Type VII Gauze Sponge, 12-Ply, 4 x 4 Inch, 16-42442, 2/Pack
Catalog No. 503042876
Encompass_Preferred
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 4 X 4 Inch Square Sterile Gauze sponges are made from 100% high-grade cotton for comfort and absorption These 4 x 4-inch USP Type VII gauze are suitable for dressing, cleaning, prepping or packing wounds They create a low amount of lint to keep the wound area clean USP Type VII gauze sponges are sterile to help ward off infection 12-ply sponges are packed in pairs in a peelable pouch for easy access 4 x 4 inches
Catalog No. 50-304-2876 Supplier Med Vet International Supplier No. 446047PK
USP Type VII Gauze Sponge McKesson Cotton 12-Ply 4 X 4 Inch Square Sterile Gauze sponges are made from 100% high-grade cotton for comfort and absorption These 4 x 4-inch USP Type VII gauze are suitable for dressing, cleaning, prepping or packing wounds They create a low amount of lint to keep the wound area clean USP Type VII gauze sponges are sterile to help ward off infection 12-ply sponges are packed in pairs in a peelable pouch for easy access 4 x 4 inches