Learn More
Med Vet International McKesson NonSterile Nonwoven Sponge, 4-Ply, 2 x 2 Inch, 92242000, 4000/Case
Catalog No. 503043576
Encompass_Preferred
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 2 X 2 Inch Square NonSterile McKesson gauze sponges have a blend of polyester and rayon which is an alternative to cotton These 4-ply sponges are suited for general cleaning Each 2-inch x 2-inch nonwoven sponge is highly absorbent They are virtually lint-free McKesson nonwoven sponges are ideal for dressing, prepping and cleaning wounds Each pack contains 200 white sponges
Catalog No. 50-304-3576 Supplier Med Vet International Supplier No. 446034CS
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 2 X 2 Inch Square NonSterile McKesson gauze sponges have a blend of polyester and rayon which is an alternative to cotton These 4-ply sponges are suited for general cleaning Each 2-inch x 2-inch nonwoven sponge is highly absorbent They are virtually lint-free McKesson nonwoven sponges are ideal for dressing, prepping and cleaning wounds Each pack contains 200 white sponges