Learn More
Med Vet International McKesson Sterile Nonwoven Sponge, 4-Ply, 4 x 4 Inch, 16-4244, 1/Pack
Catalog No. 503043113
Encompass_Preferred
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 4 X 4 Inch Square Sterile McKesson nonwoven sponges, 4 x 4 inches, are made with a polyester and rayon blend that's soft and durable Each sponge has a 4-ply thickness, good for general cleaning Nonwoven sponges are highly absorbent, making them suitable for wound dressing, prepping and cleaning The nonwoven sponge doesn't leave fibers on wounds
Catalog No. 50-304-3113 Supplier Med Vet International Supplier No. 446033PK
Nonwoven Sponge McKesson Polyester / Rayon 4-Ply 4 X 4 Inch Square Sterile McKesson nonwoven sponges, 4 x 4 inches, are made with a polyester and rayon blend that's soft and durable Each sponge has a 4-ply thickness, good for general cleaning Nonwoven sponges are highly absorbent, making them suitable for wound dressing, prepping and cleaning The nonwoven sponge doesn't leave fibers on wounds