Learn More
Med Vet International Dynarex Post Op Plus Sponge, 4" X 3", 600/Case
Catalog No. 502830326
Encompass_Preferred
Dynarex Post-Op Sponges are crafted from non-woven materials to provide improved wicking and absorbency. The Sponges are highly absorbent, sterile, and packaged in a peel-down pouch for convenient aseptic handling. These Post-Op Sponges are ideal for a wide range of surgical procedures, including dialysis. Dynarex Post-Op Sponges are premium-quality, non-woven sponges designed for increased absorbency. These sterile Sponges are ideal for a wide range of procedures, including dialysis.
Catalog No. 50-283-0326 Supplier Med Vet International Supplier No. 3355
Dynarex Post-Op Sponges are crafted from non-woven materials to provide improved wicking and absorbency. The Sponges are highly absorbent, sterile, and packaged in a peel-down pouch for convenient aseptic handling. These Post-Op Sponges are ideal for a wide range of surgical procedures, including dialysis. Dynarex Post-Op Sponges are premium-quality, non-woven sponges designed for increased absorbency. These sterile Sponges are ideal for a wide range of procedures, including dialysis.