Learn More
Med Vet International McKesson Sterile Nonwoven Sponge, 4-Ply, 4 x 4 Inch, 16-602317, 1200/Case
Catalog No. 503043560
Encompass_Preferred
Nonwoven Sponge McKesson Cotton 4-Ply 4 X 4 Inch Square Sterile McKesson gauze sponges are made of 100% cotton material to help ensure user comfort and to absorb bacteria from the wound The square shape completely covers the wound area 4-ply construction increases support, protection and absorbency They're sterile for safety and immediate usage 4 x 4 inches
Catalog No. 50-304-3560 Supplier Med Vet International Supplier No. 481054CS
Nonwoven Sponge McKesson Cotton 4-Ply 4 X 4 Inch Square Sterile McKesson gauze sponges are made of 100% cotton material to help ensure user comfort and to absorb bacteria from the wound The square shape completely covers the wound area 4-ply construction increases support, protection and absorbency They're sterile for safety and immediate usage 4 x 4 inches