Learn More
Sigma Aldrich Fine Chemicals Biosciences SpongeColR Columnar Archit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 51355EA
Encompass
SpongeCol(R) is a collagen sponge with an interpenetrating columnar porous architecture structure. SpongeCol(R) contains a unique columnar porous network which permits cells and nutrients to flow completely through the interpenetrating pores and provides an increased surface area for cell attachment growth and migration. It is composed of highly purified Type I collagen which best supports the attachment proliferation and function of cells. The collagen is lightly cross-linked for increased mechanical strength and durability for short and long term tissue culture yet is still biodegradable over the longer-term. The diameter of the pores ranges from 100 to 400 micron with the average diameter being approximately 200 microns. This product is terminally sterilized and ready-to-use.
Catalog No. 50-290-1412