Learn More
Essendant Power Sponge #3000, 2.8 x 4.5, 0.6"" Thick, Blue/Teal, 20/Carton
Catalog No. 2679735
Encompass_Preferred
Two cleaning tools in one ¿ a Scotch-Brite™ Power Pad combined with a sponge. 6-10 times less abrasive than other medium/heavy-duty commercial scouring products to reduce scratching. Antimicrobial product protection is locked-in to resist odors in the sponge.
Catalog No. 26-797-35 Supplier Essendant Supplier No. MMM3000CT
Two cleaning tools in one ¿ a Scotch-Brite™ Power Pad combined with a sponge. 6-10 times less abrasive than other medium/heavy-duty commercial scouring products to reduce scratching. Antimicrobial product protection is locked-in to resist odors in the sponge.