Learn More
Med Vet International Dynarex Advantage Surgical Sponges, 3" X 3", 12 Ply, 4000/Case
Catalog No. 502832138
Encompass_Preferred
Dynarex Advantage Surgical Sponges are highly absorbent sponges, ideal for a variety of uses, including dressing, cleaning, and prepping procedures. The Surgical Sponges are constructed from 100% cotton gauze with cut edges folded so now raw edges are exposed. Dynarex Advantage Surgical Sponges are available in sterile and non-sterile varieties. Sterile sponges are packed in peel-down pouches for convenient, aseptic handling.  Dynarex Advantage Surgical Sponges are premium-quality gauze sponges designed for cleaning, prepping, and dressing wounds. These Sponges are crafted from 100% cotton gauze for optimal absorption. 
Catalog No. 50-283-2138 Supplier Med Vet International Supplier No. 3263
Dynarex Advantage Surgical Sponges are highly absorbent sponges, ideal for a variety of uses, including dressing, cleaning, and prepping procedures. The Surgical Sponges are constructed from 100% cotton gauze with cut edges folded so now raw edges are exposed. Dynarex Advantage Surgical Sponges are available in sterile and non-sterile varieties. Sterile sponges are packed in peel-down pouches for convenient, aseptic handling.  Dynarex Advantage Surgical Sponges are premium-quality gauze sponges designed for cleaning, prepping, and dressing wounds. These Sponges are crafted from 100% cotton gauze for optimal absorption. 