Learn More
Med Vet International Dynarex Sterile Krinkle Super Sponges, 6" X 6.75", 240/Case
Catalog No. 502831005
Encompass_Preferred
Gauze rolls and sponges with great absorption and softness for packing, prepping, cleansing and bandaging. Soft, highly absorbent primary or secondary dressing Manufactured of pre-washed, fluff-dried woven gauze Krinkle weave provides cushioning support and protection for sensitive wound areas 6-Ply construction 3163 - Highly absorbent fluff-dried gauze with unique construction and diamond fold offers cushioning protection for the wound site; 12-Ply construction
Catalog No. 50-283-1005 Supplier Med Vet International Supplier No. 3163
Gauze rolls and sponges with great absorption and softness for packing, prepping, cleansing and bandaging. Soft, highly absorbent primary or secondary dressing Manufactured of pre-washed, fluff-dried woven gauze Krinkle weave provides cushioning support and protection for sensitive wound areas 6-Ply construction 3163 - Highly absorbent fluff-dried gauze with unique construction and diamond fold offers cushioning protection for the wound site; 12-Ply construction