Learn More
SPT LABTECH INC Spool of 18500 Gamma Ray Sterilised Pipettes 4.5mm Pitch HV
Catalog No. NC3245866
Encompass
Disposable gamma ray sterilized positive-displacement pipettes give superior accuracy for a wide range of liquid viscosities. Volume range of 500 nL to 5 μL, suitable for addressing 384- and 1,536-well plates.
Catalog No. NC3245866 Supplier SPT LABTECH INC Supplier No. 415003033
Disposable gamma ray sterilized positive-displacement pipettes give superior accuracy for a wide range of liquid viscosities. Volume range of 500 nL to 5 μL, suitable for addressing 384- and 1,536-well plates.