Learn More
Chemglass Life Sciences Spool Racks, Mobile, holds up to 6 each 13 spools
SDP
Catalog No. 501311194
Encompass_Preferred
Spool Rack, Mobile. Rack is on casters making it easy to move from lab to lab. It will hold up to 6 each 13 spools. With the rack being only 17 wide it can fit into tight spaces between lab benches and hoods. Constructed of powder coated aluminum making the unit light and resistant to most lab environments. Rack is 35 Long x 17 Wide x 52 tall.
Catalog No. 50-131-1194 Supplier Chemglass Life Sciences Supplier No. CG1997R10
Spool Rack, Mobile. Rack is on casters making it easy to move from lab to lab. It will hold up to 6 each 13 spools. With the rack being only 17 wide it can fit into tight spaces between lab benches and hoods. Constructed of powder coated aluminum making the unit light and resistant to most lab environments. Rack is 35 Long x 17 Wide x 52 tall.