Learn More
SPT LABTECH INC Spool Of 26000 Non-Sterile Pipettes 9mm Pitch
Catalog No. NC0012189
Encompass
Disposable positive-displacement pipettes give superior accuracy for a wide range of liquid viscosities. Volume range of 25 nL to 1.2 μL, suitable for addressing 96-, 384- and 1,536-well plates.
Catalog No. NC0012189 Supplier SPT LABTECH INC Supplier No. 415003020
Disposable positive-displacement pipettes give superior accuracy for a wide range of liquid viscosities. Volume range of 25 nL to 1.2 μL, suitable for addressing 96-, 384- and 1,536-well plates.