Learn More
BURKLE INC Sampling spoon curved, l.h., 0.34 oz. (10 ml), Bio-PE
Catalog No. 503081180
Encompass_Preferred
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
Catalog No. 50-308-1180 Supplier BURKLE INC Supplier No. 53790023
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.