Learn More
BURKLE INC Detectable spoon curved, l. h., 0.34 oz. (10 ml), PS, sterile
Catalog No. 503081190
Encompass_Preferred
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The spoon with curved spoon bowl is suitable for a wide variety of media, e.g. powders, granulates, pastes or liquids. The angled spoon shape makes it easy to reach the corners of a container. The long-handled plastic scoop allows larger containers to be emptied easily and can also reach places that are difficult to access, e.g. narrow-necked canisters.
Catalog No. 50-308-1190 Supplier BURKLE INC Supplier No. 53785012
Add to Cart
Add to Cart
In addition to the blue color, these samplers have a special additive in the material that makes them visible in metal detectors or by X-ray. Although they are made of plastic, these products can therefore be sorted out using the usual systems for checking for foreign bodies.
The spoon with curved spoon bowl is suitable for a wide variety of media, e.g. powders, granulates, pastes or liquids. The angled spoon shape makes it easy to reach the corners of a container. The long-handled plastic scoop allows larger containers to be emptied easily and can also reach places that are difficult to access, e.g. narrow-necked canisters.