Learn More
BURKLE INC Food spoons, blue, PS, sterile,0.08 oz. (2,5 ml)
Supplier:  BURKLE INC 53783011
Encompass_Preferred
The SteriPlast spoon for foodstuffs, made of polystyrene, is produced in our class 7 clean room (10,000), individually packaged for disposable use and then sterilized by gamma rays.
The spoon for foodstuffs is available in two sizes, 0.08 oz. (2.5 ml) (0.08 oz.) (similar to a teaspoon) and 10 ml (0.34 oz.) (similar to a tablespoon), and is ideal for sampling powders, granulates, pastes and fluids.
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects. This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry.
Catalog No. 50-308-1281