Learn More
BURKLE INC Sampling spoon, long handle, 0.17 oz. (5 ml), PS,white,sterile
Catalog No. 503081187
Encompass_Preferred
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
Catalog No. 50-308-1187 Supplier BURKLE INC Supplier No. 53785002
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.