Learn More
Foxx Life Sciences EZBio Stainless Steel Spoon-Spatula Polished with Steel Handle 304 SS, 180mm, 5/PK
Catalog No. NC1795131
Encompass_Preferred
Foxx Life Sciences Stainless Steel Spoon-Spatulas are designed for ease of use and versatility. This double-sided, multifunctional device features a spoon on one side and a spatula on the other. These ideal products make it possible to support numerous processes, such as sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These double-sided spoons are available in sizes ranging from 120mm to 300mm.304 Stainless Steel High-gloss polishedDouble-sided functionalityEasy to clean hygienic design
Catalog No. NC1795131 Supplier Foxx Life Sciences Supplier No. 25C3618FLS
Foxx Life Sciences Stainless Steel Spoon-Spatulas are designed for ease of use and versatility. This double-sided, multifunctional device features a spoon on one side and a spatula on the other. These ideal products make it possible to support numerous processes, such as sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These double-sided spoons are available in sizes ranging from 120mm to 300mm.304 Stainless Steel High-gloss polishedDouble-sided functionalityEasy to clean hygienic design