Learn More
BURKLE INC Spoon spatula, LaboPlast, PS, 0.02 oz. (0,5 ml)/ 0.67 in (17 mm)
Catalog No. 503081316
Encompass_Preferred
The LaboPlast spoon spatula is a dual-ended tool featuring a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67 in.) spatula blade, with a total length of 178 mm (7.01 in.).
Manufactured in a cleanroom environment, this disposable spatula is made from durable PS material that is chemical-resistant and corrosion-proof, ideal for laboratory, medical, and cosmetic applications requiring high hygiene standards.
Choose the LaboPlast spoon spatula for a reliable, cleanroom-produced, and convenient single-use tool.
Catalog No. 50-308-1316 Supplier BURKLE INC Supplier No. 53780031
The LaboPlast spoon spatula is a dual-ended tool featuring a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67 in.) spatula blade, with a total length of 178 mm (7.01 in.).
Manufactured in a cleanroom environment, this disposable spatula is made from durable PS material that is chemical-resistant and corrosion-proof, ideal for laboratory, medical, and cosmetic applications requiring high hygiene standards.
Choose the LaboPlast spoon spatula for a reliable, cleanroom-produced, and convenient single-use tool.