Learn More
BURKLE INC Spoon spatula, SteriPlast, PS, sterile, 0.02 oz. (0,5 ml)/ 0.67 in (17 mm)
Catalog No. 503081284
Encompass_Preferred
The SteriPlast spoon spatula is a versatile, double-ended tool featuring a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67 in.) spatula blade, with a total length of 178 mm (7.01 in.).
Designed for single-use applications, this sterile spatula is made from durable PS material, ensuring chemical resistance and corrosion-proof performance. Ideal for precise measuring and mixing in labs, medical, and cosmetic settings.
Choose the SteriPlast spoon spatula for a reliable, hygienic, and convenient disposable solution.
Catalog No. 50-308-1284 Supplier BURKLE INC Supplier No. 53781031
The SteriPlast spoon spatula is a versatile, double-ended tool featuring a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67 in.) spatula blade, with a total length of 178 mm (7.01 in.).
Designed for single-use applications, this sterile spatula is made from durable PS material, ensuring chemical resistance and corrosion-proof performance. Ideal for precise measuring and mixing in labs, medical, and cosmetic settings.
Choose the SteriPlast spoon spatula for a reliable, hygienic, and convenient disposable solution.