Learn More
BURKLE INC Sampling spoon curved, l.h., 0.34 oz. (10 ml),PS,white,sterile
Catalog No. 503081366
Encompass_Preferred
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
Catalog No. 50-308-1366 Supplier BURKLE INC Supplier No. 53785010
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.