Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Spor-Klenz™ Concentrate Cold Sterilant

Catalog No. 50465417 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
50-465-417 1 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-465-417 Supplier STERIS Supplier No. 652026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An easy-to-dilute, sporicidal disinfectant that offers a low toxicity profile and requires no activation

  • Specifically formulated for use in the sterilization and disinfection of hard environmental surfaces
  • A stabilized blend of peracetic acid, hydrogen peroxide, and acetic acid
  • Versatility of use as a sterilant, sporicide, disinfectant, or sanitizer
  • Microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC methods
  • For use in Industrial Laboratories, Pharmaceutical, Medical Device, Biotech, and Cosmetic manufacturing operations
  • Can be applied manually, sprayed, or fogged
  • No activation required
  • One quart makes 25 gallons of use solution at a dilution of 1:99
  • Use-dilution can be used for up to 7 days to reduce waste
  • Each lot is fully traceable and documented by a Certificate of Analysis that includes filtration and USP sterility

Specifications

Product Type Sterilant Disinfectant Sporicide
Form Concentrate
Sterility Non-sterile
Composition A Stabilized Blend of Peracetic Acid, Hydrogen Peroxide, and Acetic Acid
Filtered Filtered
For Use With (Application) Sterilizing and Disinfecting Hard Environmental Surfaces
Quantity 4/Cs.
Volume (English) 1 qt.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.