Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Spor-Klenz™ Ready-To-Use Disinfectant Wipes

Catalog No. 19415145 Shop All STERIS Corporation Products
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-415-145 9 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-415-145 Supplier STERIS Supplier No. 652915
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spor-Klenz™ RTU Disinfectant wipes kill a broad spectrum of common bacteria, fungi, and viruses.

  • Innovative system combines woven polyester wipes with sealed edges packaged with a separate internal pouch of Spor-Klenz RTU formula.
  • Internal pouch allows for maximum chemical stability while offering a
    simple, convenient and consistent method to saturate the wipes.
  • By simply rolling the package of wipes, the internal pouch of liquid opens and quickly saturates the wipes.
  • Unique packaging system utilizes the same Spor-Klenz RTU formula used for mopping or spraying and creates saturated wipes with the least amount of operator effort.
  • Extensive documentation package to meet your validation objectives and are supported by the STERIS Technical Services team for individualized application and validation assistance.
  • Applications: Surface disinfection and microbial control of:
    • Countertops
    • BSL hoods
    • Carts and other equipment

Specifications

Color White
Dimensions (L x W) 9 x 9 in.
Format Double bagged and gamma irradiated
Packaging Quantity 12 Pouches/Cs. (180 Wipes/Cs.)
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.