Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Spor-Klenz™ Ready-To-Use Sterilant/Disinfectant

Catalog No. 19415151 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (Metric):
850 mL
3200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-415-151 850 mL
04-324-20 3200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-415-151 Supplier STERIS Supplier No. 6525M2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Sterilant/Disinfectant only. Sprayer is sold separately (see Cat No. 6526Q8).

Provides consistent microbial control, no mixing or activation is required. Spray or apply manually straight out of the container.

  • Specifically formulated for the sterilization and disinfection of hard environmental surfaces
  • A stabilized blend of hydrogen peroxide, peracetic, and acetic acids
  • Provides effective microbial control, including spores
  • Filtered to 0.22 μm
  • Offers a low toxicity profile that requires no mixing or activation
  • Reusable for up to 14 days to reduce waste
  • For use in Pharmaceutical, Medical Device, Biotech, and Cosmetic manufacturing facilities
  • All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD)
  • Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility
  • NOTE: Sprayer is sold separately (see Cat No. 6526Q8)   

Specifications

Product Type Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner
Certifications/Compliance European Union Biocidal Products Directive (BPD)
Composition Stabilized Blend of Hydrogen Peroxide, Peracetic, and Acetic Acids
Container Type Plastic Bottle (sprayer sold separately.)
Filtered Filtered
For Use With (Application) Sterilization and Disinfection of Hard Environmental Surfaces
Quantity 4/Cs.
Volume (Metric) 850 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.