Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ Spor-Klenz RTU Sterilant

Catalog No. 19999572 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
50 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-999-572 50 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-999-572 Supplier STERIS™ Supplier No. 6525P4
Add to Cart
Add to Cart

Spor-Klenz Ready-to-Use Sterilant is a cleanroom disinfectant, sterilant and sporicidal cleaner that can be sprayed or applied manually straight out of the container. There’s no mixing or activation required, providing consistent microbial control.

STERIS' Spor-Klenz Ready-To-Use Cold Sterilant is a fast acting, liquid cold sterilant/disinfectant, filtered to 0.22 micron and specifically formulated for use in the sterilization and disinfection of hard environmental surfaces in Pharmaceutical, Medical Device, Biotech and Cosmetic manufacturing facilities. This product is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic that provides fast, effective microbial control, including spores. All of the active ingredients are notified according to the European Union Biocidal Products Directive (BPD). Spor-Klenz Ready-To-Use Sterilant offers a low toxicity profile, requires no mixing or activation and is reusable for up to 14 days which reduces waste. Each lot is fully traceable and documented by a Certificate of Analysis including filtration and USP sterility.

HOW SPOR-KLENZ READY-TO-USE COLD STERILANT WORKS

STERIS' Spor-Klenz Ready-To-Use Sterilant is a liquid sporicide specifically formulated for sterilization and disinfection of hard environmental surfaces. It can be used straight out of the container and be applied manually, sprayed or fogged.

WHY SPOR-KLENZ READY-TO-USE COLD STERILANT?

  • Ease of Use - Consistent dilution every time because no mixing or activation is required.
  • Efficacy - Microbial control against bacteria, fungi, viruses, and bacteria spores proven using AOAC and EN protocols.
  • Safety - Low toxicity profile to support worker safety.
  • Convenience - Excellent material compatibility allows use on most environmental surfaces.
  • Flexibility - Versatility of use as a sterilant, sporicide, disinfectant or sanitizer.

Specifications

Product Type Cleanroom Disinfectant, Sterilant, and Sporicidal Cleaner
Form Clear Liquid
pH 1.8 (Undiluted)
Scent Unscented
Sterility Non-sterile
Composition Peracetic Acid, Hydrogen Peroxide, Acetic Acid
Container Type Drum
Volume (English) 50 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.