Learn More
SPS Medical SPOR VIEW STARTER KIT
Catalog No. NC9865958
Encompass_Preferred
SporView Self-Contained Biological Indicators are approved for use in steam sterilization processes. Used for immediate release of routine or immediate use non-implant loads. Starter kit comes with an incubator, record notebook and 25 BI's.
Catalog No. NC9865958 Supplier SPS Medical Supplier No. SK116
SporView Self-Contained Biological Indicators are approved for use in steam sterilization processes. Used for immediate release of routine or immediate use non-implant loads. Starter kit comes with an incubator, record notebook and 25 BI's.