Learn More
MSC EvaClean Disinfectant Tablets, 13.1 g, Sporicidal, 100 Count
Catalog No. 502819455
Encompass
PurOne Effervescent Cleaner & Disinfectant Tablets are multi-purpose, hospital-grade solutions that activate with tap water to clean and disinfect. They are designed to eliminate the need for multiple chemicals, effectively killing bacteria, pathogens, and biofilm in as little as 4 minutes.
  • Kills C. diff and COVID-19 in 4 minutes
  • Kills biofilm bacteria and C. auris
  • Sporicidal grade disinfectant
  • Neutral pH (6.5–7.0)
  • Contains 48.21% sodium dichloro-s-triazinetrione
  • EPA-registered (71847-7-91524)
  • Approved for use against emerging pathogens (EPA List K)
  • Complies with OSHA bloodborne pathogen standard
Catalog No. 50-281-9455 Supplier MSC Supplier No. 11669041
PurOne Effervescent Cleaner & Disinfectant Tablets are multi-purpose, hospital-grade solutions that activate with tap water to clean and disinfect. They are designed to eliminate the need for multiple chemicals, effectively killing bacteria, pathogens, and biofilm in as little as 4 minutes.
  • Kills C. diff and COVID-19 in 4 minutes
  • Kills biofilm bacteria and C. auris
  • Sporicidal grade disinfectant
  • Neutral pH (6.5–7.0)
  • Contains 48.21% sodium dichloro-s-triazinetrione
  • EPA-registered (71847-7-91524)
  • Approved for use against emerging pathogens (EPA List K)
  • Complies with OSHA bloodborne pathogen standard