Learn More
Sigma Aldrich Fine Chemicals Biosciences Schaeffer and Fulton Spore Stain Kit, 1 kit
Catalog No. NC1726367
Encompass
In Schaeffer-Fulton`s method, a primary stain-malachite green is forced into the spore by steaming the bacterial emulsion. Malachite green is water soluble and has a low affinity for cellular material, so vegetative cells may be decolorized with water. Vegetative cells are then counterstained with safarin.
Catalog No. NC1726367 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 045511KTF
In Schaeffer-Fulton`s method, a primary stain-malachite green is forced into the spore by steaming the bacterial emulsion. Malachite green is water soluble and has a low affinity for cellular material, so vegetative cells may be decolorized with water. Vegetative cells are then counterstained with safarin.