Learn More
Weber Scientific SPORE STRIP BIO INDCTR 100/PK
SDP
Catalog No. NC0593545
Encompass
Used to validate and routinely monitor the adequacy of steam, ethylene oxide, dry heat and chemical vapor sterilization.
Catalog No. NC0593545 Supplier Weber Scientific Supplier No. 305796
Used to validate and routinely monitor the adequacy of steam, ethylene oxide, dry heat and chemical vapor sterilization.