Learn More
MESA LABS BOZEMAN MFG FACILITY SPORE STRIPS 100/PK
SDP
Catalog No. NC9688500
Encompass
SGMStrip, CONVENTIONAL SPORE STRIP BIOLOGICAL INDICATORS, Availability:Geobacillus stearothermophilus, Process:steam, Population:106, Incubation Time:7 days, Incubation Temp:55 - 60 deg C
Catalog No. NC9688500 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. SGMS/6
SGMStrip, CONVENTIONAL SPORE STRIP BIOLOGICAL INDICATORS, Availability:Geobacillus stearothermophilus, Process:steam, Population:106, Incubation Time:7 days, Incubation Temp:55 - 60 deg C