Learn More
CROSSTEX INTERNATIONAL Spore Suspensions, 10^2, 10 mL
Catalog No. NC1420492
Encompass_Preferred
Crosstex manufactures a wide range of Spore Suspensions utilized in Ethylene Oxide (EO) and Dry Heat sterilization processes. The Spore Suspensions are pure suspensions of spores with known resistance characteristics and a variety of population levels standardized per 0.1 mL. The Spore Suspensions allow for direct inoculation of products, typically to verify sterility of devices where a traditional biological indicator spore strip cannot be used.
Catalog No. NC1420492 Supplier CROSSTEX INTERNATIONAL Supplier No. VBA102
Crosstex manufactures a wide range of Spore Suspensions utilized in Ethylene Oxide (EO) and Dry Heat sterilization processes. The Spore Suspensions are pure suspensions of spores with known resistance characteristics and a variety of population levels standardized per 0.1 mL. The Spore Suspensions allow for direct inoculation of products, typically to verify sterility of devices where a traditional biological indicator spore strip cannot be used.