Learn More
SPS Medical SPORE VIEW CULTURE SET PK/20
Catalog No. NC9494532
Encompass
SporView Culture Set,20/box,Description:A convenient BI monitoring system designed to provide sterilization verification right in the department where processing is done,Function:For use in Steam,EO gas
Catalog No. NC9494532 Supplier SPS Medical Supplier No. CS020
SporView Culture Set,20/box,Description:A convenient BI monitoring system designed to provide sterilization verification right in the department where processing is done,Function:For use in Steam,EO gas