Learn More
CROSSTEX INTERNATIONAL SPORE WIRE STERILIZATION 100PK
Catalog No. NC1772358
Encompass_Preferred
spore wire inoculated carriers for monitoring steam sterilization processes,braided steel wires each 40mm length and <2mm dia,bulked, geobacillus stearothermophilus cell line 7953
Catalog No. NC1772358 Supplier CROSSTEX INTERNATIONAL Supplier No. NMSWS06
spore wire inoculated carriers for monitoring steam sterilization processes,braided steel wires each 40mm length and <2mm dia,bulked, geobacillus stearothermophilus cell line 7953