Learn More
ATCC Sporosarcina pasteurii
Catalog No. 502384757
Encompass
Sporosarcina pasteurii is a bacterial type strain and is known to produce the urease urea amidolyase.
Catalog No. 50-238-4757 Supplier ATCC Supplier No. 11859
Sporosarcina pasteurii is a bacterial type strain and is known to produce the urease urea amidolyase.