Learn More
Qc Supply LLC SPORTSMAN 1502 EGG INCUBATOR
SDP
Catalog No. NC0434220
Encompass
  • Digital thermostat for precise temperature control
  • Integrated LCD for temperature and humidity monitoring
  • Automatic egg turning with 3 racks
  • Constructed with high-strength plastic board for durability
  • Temperature range of 60 to 103° F
  • Input power of 110-120 VAC, 50/60 Hz
  • Power consumption of 325 watts
  • TUV safety listed for quality assurance
  • Dimensions: 30 1/4 in x 15 3/4 in x 31 3/4 in
  • Weight options: 75 lb, 90 lb, or 96 lb
  • Capacity for 288 chicken eggs
  • Capacity for 1368 quail eggs
  • Capacity for 354 pheasant eggs
  • Capacity for 198 turkey/duck eggs
  • Can accommodate 36-45 large eggs (goose, peafowl, emu)
  • Hatching drawer holds approximately 72 medium eggs
  • Includes clear viewing door for easy monitoring
  • Comes with various essential components for egg incubation
Catalog No. NC0434220 Supplier Qc Supply LLC Supplier No. 420390
  • Digital thermostat for precise temperature control
  • Integrated LCD for temperature and humidity monitoring
  • Automatic egg turning with 3 racks
  • Constructed with high-strength plastic board for durability
  • Temperature range of 60 to 103° F
  • Input power of 110-120 VAC, 50/60 Hz
  • Power consumption of 325 watts
  • TUV safety listed for quality assurance
  • Dimensions: 30 1/4 in x 15 3/4 in x 31 3/4 in
  • Weight options: 75 lb, 90 lb, or 96 lb
  • Capacity for 288 chicken eggs
  • Capacity for 1368 quail eggs
  • Capacity for 354 pheasant eggs
  • Capacity for 198 turkey/duck eggs
  • Can accommodate 36-45 large eggs (goose, peafowl, emu)
  • Hatching drawer holds approximately 72 medium eggs
  • Includes clear viewing door for easy monitoring
  • Comes with various essential components for egg incubation