Learn More
Proteintech Group Inc Spot-Label Alexa Fluor 647
Supplier: Proteintech Group Inc EBAF64710
anti-Spot-Tag VHH/ Nanobody conjugated to fluorescent dye for immunofluorescence, microscopy, and immunoblotting of Spot-tagged proteins.