Learn More
Invitrogen™ SPoT-Light Tissue Pretreatment Kit
Description
For Enzyme and Heat Pretreatment of FFPE Tissue prior to CISH.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). Shelf life: 18 months from date of manufacture |
| Type | Tissue Pretreatment Kit |
| Product Line | CISH, SPOT-Light |
| Quantity | 1 Kit |
| Reagent Type | Antigen Retrieval Reagent |
| Shipping Condition | Wet Ice |
Safety and Handling
For Laboratory Use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.