Learn More
Proteintech Group Inc Spot-Trap Magnetic Particles M-270
SDP
Supplier:  Proteintech Group Inc ETD10
Encompass
Spot-Trap Magnetic Particles M-270 are affinity beads for immunoprecipitation (IP) of Spot-tagged proteins It comprises an anti-Spot-Tag VHH/ Nanobody coupled to Magnetic Particles M-270.
Catalog No. 50-225-8183