Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ SpotDrops, Catalase Reagent, Hydrogen Peroxide
SDP

Catalog No. 23001803 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
23-001-803 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-803 Supplier Hardy Diagnostics™ Supplier No. Z62
Add to Cart
Add to Cart
This item is not returnable. View return policy

Useful in the presumptive identification and differentiation of many bacteria. Beta-hemolytic organisms, such as Streptococcus species Staphylococcus species, and Listeria species can be differentiated by their catalase reaction.

  • Hydrogen peroxide 3%, (H2O2), catalase positive organisms (such as Staphylococcus) will evolve gas bubbles in the presence of H2O2

Specifications

Quantity 15 mL
Color Colorless
Volume (Metric) 15 mL
Detection Method Bubbles Indicate Positive Test
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.