Learn More
Maine Molecular Quality Controls Inc SPOTFIRE RSP Pos & Neg Controls
SDP
Catalog No. NC3886279
Encompass
The SPOTFIRE RSP Pos & Neg Controls kit is intended for use (as applicable) as an external positive and negative assayed quality control to monitor performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Bordetella parapertussis, Bordetella pertussis, Chlamydia pneumoniae, Mycoplasma pneumoniae, Streptococcus dysgalactiae (Group C/G Strep), Streptococcus pyogenes (Group A Strep), Adenovirus, Coronavirus SARS-CoV-2, Coronavirus (seasonal), Human Metapneumovirus, Human Rhinovirus/Enterovirus, Influenza A Subtype H1-2009, Influenza A Subtype H3, Influenza B, Parainfluenza Virus and Respiratory Syncytial Virus using the BIOFIRE SPOTFIRE Respiratory (R) Panel, BIOFIRE SPOTFIRE Respiratory (R) Panel Mini, BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel and BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel Mini on the BIOFIRE SPOTFIRE System.
Catalog No. NC3886279 Supplier Maine Molecular Quality Controls Inc Supplier No. M425
The SPOTFIRE RSP Pos & Neg Controls kit is intended for use (as applicable) as an external positive and negative assayed quality control to monitor performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of Bordetella parapertussis, Bordetella pertussis, Chlamydia pneumoniae, Mycoplasma pneumoniae, Streptococcus dysgalactiae (Group C/G Strep), Streptococcus pyogenes (Group A Strep), Adenovirus, Coronavirus SARS-CoV-2, Coronavirus (seasonal), Human Metapneumovirus, Human Rhinovirus/Enterovirus, Influenza A Subtype H1-2009, Influenza A Subtype H3, Influenza B, Parainfluenza Virus and Respiratory Syncytial Virus using the BIOFIRE SPOTFIRE Respiratory (R) Panel, BIOFIRE SPOTFIRE Respiratory (R) Panel Mini, BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel and BIOFIRE SPOTFIRE Respiratory/Sore Throat (R/ST) Panel Mini on the BIOFIRE SPOTFIRE System.