Learn More
BRUKER CELLULAR ANALYSIS INC SPOTLIGHT HUMAN FC REAGENT
SDP
Catalog No. NC2747955
Encompass
  • Highly concentrated assay reagent for advanced cell line development workflows.
  • Specifically binds to the Fc region of human antibodies.
  • Non-destructive and animal-component-free fluorescent reagent.
  • Designed for efficient screening of monoclonal or bispecific antibody productivity and quality.
  • Compatible with Bruker's Beacon and Beacon Select™ optofluidic systems.
  • Supports Opto CLD 1.0 and 2.0 workflows.
  • Volume: 200 µL.
  • Uses per unit: 1-2 workflows.
  • Storage temperature: 4°C.
  • Regulatory status: For research use only.
Catalog No. NC2747955 Supplier BRUKER CELLULAR ANALYSIS INC Supplier No. 52000024
Add to Cart
Add to Cart
Request a Quote
  • Highly concentrated assay reagent for advanced cell line development workflows.
  • Specifically binds to the Fc region of human antibodies.
  • Non-destructive and animal-component-free fluorescent reagent.
  • Designed for efficient screening of monoclonal or bispecific antibody productivity and quality.
  • Compatible with Bruker's Beacon and Beacon Select™ optofluidic systems.
  • Supports Opto CLD 1.0 and 2.0 workflows.
  • Volume: 200 µL.
  • Uses per unit: 1-2 workflows.
  • Storage temperature: 4°C.
  • Regulatory status: For research use only.